Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Corning® Minimum Essential Medium (Mod.) (MEM), Corning

Corning® Minimum Essential Medium (Mod.) (MEM), Corning

MEM, Corning® 10 L MEM (Minimum Essential Medium), powder [+] Earles salts, L-glutamine, NEAA [-] sodium bicarbonate, Powder
Catalog # 45000-594
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Catalog # 45000-594
Supplier:  
CAS Number:  
Corning® Minimum Essential Medium (Mod.) (MEM), Corning
Corning® Minimum Essential Medium (Mod.) (MEM), Corning

MEM, Corning® 10 L MEM (Minimum Essential Medium), powder [+] Earles salts, L-glutamine, NEAA [-] sodium bicarbonate, Powder

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
MEDIUM MEME W/ES/NEAA/GLN W/O NAHCO3 5L Catalog #95016-562

Product 95016-562 is a replacement for 45000-594 but varies by Brand, Packaging or Quantity.

MEDIUM MEME PWD CELLGRO W/O SOD PYR 50L Catalog #45000-596

Product 45000-596 is a replacement for 45000-594 but varies by Brand, Packaging or Quantity.

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Minimum Essential Medium (Mod.)(EMEM) based on Basal Medium Eagle (BME) formulation to aid cell propagation.

  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment

It is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts. This formulation contains Earle's salts and with or without L-glutamine.

Specifications

Specifications

  • Media Format:
    Powder
  • Description:
    MEM, Corning® 10 L MEM (Minimum Essential Medium), powder [+] Earles salts, L-glutamine, NEAA [-] sodium bicarbonate
  • Cat. No.:
    45000-594
  • Storage Temperature:
    Refrigerate

Specifications