Orders LinkContactUsLinkComponent
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
MEM, Corning® 500 ml MEM (Minimum Essential Medium) alpha medium [+] Earles salts [-] ribonucleosides, deoxyribonucleosides, L-glutamine, Liquid, 500 ml
  45000-302
 :   Corning
 :  15-012-CV
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
  45000-302
 :  Corning
 :  
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
Corning® MEM (Minimum Essential Medium) Alpha Medium, Corning
MEM, Corning® 500 ml MEM (Minimum Essential Medium) alpha medium [+] Earles salts [-] ribonucleosides, deoxyribonucleosides, L-glutamine, Liquid, 500 ml
 :  
$170.00
Pack of 6 (500ml)
Minimum Essential Medium (MEM) is a common cell culture medium developed from early work using Basal Medium Eagle (BME) with normal mammalian fibroblasts and certain subtypes of HeLa cells.
  • Low endotoxin - < 0.25EU/mL
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
MEM is modified with higher concentrations of amino acids to more closely approximate the protein composition of mammalian cells. When supplemented with serum, MEM has been used for cultivation of a wide variety of cells grown in monolayers, such as fibroblasts.
 : Triple 0.1 µm sterilized.
  • Media Format:
    Liquid
  • Description:
    MEM, Corning® 500 ml MEM (Minimum Essential Medium) alpha medium [+] Earles salts [-] ribonucleosides, deoxyribonucleosides, L-glutamine
  • Size:
    500 ml
  • Cat. No.:
    45000-302
  • Storage Temperature:
    2...8 °C