Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PRIMER PRIS R MED 7701 CS10
Catalog # 37001-750
Supplier:
Loctite
SDS
Expand image
PRIMER PRIS R MED 7701 CS10
Catalog # 37001-750
Supplier:
Loctite
Supplier Number:
135449
SDS
Related Information
Documentation
SDS
ghs_19886
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation