Orders LinkContactUsLinkComponent
Disposable Inoculation Needle, Plastic
Disposable Inoculation Needle, Plastic
Inoculating Needle, Violet, 20/Peel Pouch
Catalog # 30002-108
Supplier:   Copan Diagnostics
Supplier #:  176CS20
Disposable Inoculation Needle, Plastic
Catalog # 30002-108
Supplier:  Copan Diagnostics
Disposable Inoculation Needle, Plastic
Disposable Inoculation Needle, Plastic
Inoculating Needle, Violet, 20/Peel Pouch
Price
$309.02
Log in for availability
Case of 1,000
Product Details & Documents
COPAN Plastic Inoculation Needles have an ultra-smooth surface to avoid gouging agar
Plastic needles are free of lubricants, oils, and electrostatic charges.
Documents
Specifications
  • Color:
    Violet
  • Description:
    Inoculating Needle
  • Packaging:
    20/Peel Pouch