Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DECON-CYCLE 16OZ .125 N/S
Catalog # 14003-678
Supplier:
Veltek Associates
SDS
Expand image
DECON-CYCLE 16OZ .125 N/S
Catalog # 14003-678
Supplier:
Veltek Associates
Supplier Number:
DCY2-07-16Z-02
SDS
Related Information
Documentation
SDS
ghs_DCY-04
Documentation