Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GAS.75%VOL(50%LEL)PENT 20.9%O2
Catalog # 13453-516
Supplier:
Draeger
SDS
Expand image
GAS.75%VOL(50%LEL)PENT 20.9%O2
Catalog # 13453-516
Supplier:
Draeger
Supplier Number:
4510057
SDS
Related Information
Documentation
SDS
ghs_4510057.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation