Technical Buffer Solutions, Sachets
Product Details & Documents
NIST-traceable buffer for pH electrode calibration and verification.
- NIST-traceable
- Ensure freshness
- Improve data
Also available in multiple packaging options.
Mettler Toledo buffer sachets ensure a fresh portion of buffer is used for each calibration, improving your pH data.
Documents
Specifications
Specifications
- Pack type:Sachet, red
- Size:30×20 ml
- pH:4.01
- pH accuracy:±0.02