$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
GLOVE STRL DURAPRENE 8.5 PK80
PH2D72A6I
:
Cardinal Health
GLOVE STRL DURAPRENE 8.5 PK80
PH2D72A6I
:
Cardinal Health
:
2D72A6I