Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CLASS 4 3KG INDIVIDUAL SATINW
Catalog # AP400002-168
Supplier:
Rice Lake Weighing
Expand image
CLASS 4 3KG INDIVIDUAL SATINW
Catalog # AP400002-168
Supplier:
Rice Lake Weighing
Supplier Number:
13278