Chlorophenol red
Bottle, 25 g
Supplier: Thermo Scientific Chemicals
Supplier #: B21623-14
Jump to:
Product Details & Documents
CAS No.: 4430-20-0 Molecular Formula: C19H12Cl2O5S Formula Weight: 432.28 Beilstein Reference: 354053 MDL No.: MFCD00005877
Synonyms: 3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
- CAS Number:4430-20-0
- Formula:C₁₉H₁₂Cl₂O₅S
- MDL Number:MFCD00005877
- Molecular Weight:423.27 g/mol
- Melting Pt:∼261 °C
- Storage Temperature:Ambient
- EINECS:224-619-2
Documents
Specifications
Specifications
- Pack type:Bottle
- Size:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



