Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lead(II) acetate basic

Lead(II) acetate basic

bottle, 2500 g
Catalog # AAAA16763-0E
Health
Environment
Supplier #:  A16763-0E
Lead(II) acetate basic
Catalog # AAAA16763-0E
CAS Number:  1335-32-6
Lead(II) acetate basic

bottle, 2500 g

CAS Number:  1335-32-6
Price
$281.06
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

CAS No.: 1335-32-6 Molecular Formula: C4H6O4Pb.2Pb(OH)2 Formula Weight: 807.72 Merck Reference: 14,5419 UN No.: 1616 MDL No.: MFCD00013041

Synonyms:  Horne′s compound, Lead(II) hydroxide acetate, Acetic acid lead (II) salt basic, Lead subacetate
  • CAS Number:1335-32-6
  • Formula:(CH₃COO)₂Pb·2Pb(OH)₂
  • MDL Number:MFCD00011154
  • Molecular Weight:807.72 g/mol
  • Merck Index:14,5419
  • Storage Temperature:Ambient
  • EINECS:215-630-3
  • UN:1616
  • ADR:6.1,III

Documents

Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    2500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.