Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

2-Ethoxyethanol 99%

bottle, 500 ml
Catalog # AAAA16100-AP
Flammable
Toxic
Health
 :  A16100-AP
2-Ethoxyethanol 99%
Catalog # AAAA16100-AP
CAS Number:  110-80-5
2-Ethoxyethanol 99%
2-Ethoxyethanol 99%

bottle, 500 ml

CAS Number:  110-80-5
Price
$55.78
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A widely used solvent for nitrocelluloses and dyes

Purity: 99% CAS No.: 110-80-5 Molecular Formula: C4H10O2 Formula Weight: 90.12 Flash Point: 44°(111°F) Refractive Index: 1.4085 Odor: Ether-like Melting Point: -90° Boiling Point: 134-135° Beilstein Reference: 1098271 Merck Reference: 14,3750 UN No.: 1171 MDL No.: MFCD00002869

Synonyms:  Ethyl glycol, Ethylene glycol monoethyl ether, Cellosolve
  • CAS Number:110-80-5
  • Formula:C₂H₅OCH₂CH₂OH
  • MDL Number:MFCD00002869
  • Molecular Weight:90.12 g/mol
  • Merck Index:14,3750
  • Boiling Pt:134…135 °C
  • Melting Pt:–90 °C
  • Density:0.931
  • Flash Pt:44 °C (111 °F)
  • Storage Temperature:Ambient
  • EINECS:203-804-1
  • UN:1171
  • ADR:3,III
  • REACH:01-2119560582-38
Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.