Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
L(+)-Ascorbic acid 99+%

L(+)-Ascorbic acid 99+%

bottle, 50 g
Catalog # AAAA15613-18
L(+)-Ascorbic acid 99+%
Catalog # AAAA15613-18
Supplier:  
CAS Number:  50-81-7
L(+)-Ascorbic acid 99+%
L(+)-Ascorbic acid 99+%

bottle, 50 g

CAS Number:  50-81-7
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
L-ASCORBIC ACID 25GM Catalog #TCA0537-025G

Product TCA0537-025G is a replacement for AAAA15613-18 and has the same packaging and specifications, but may be a different quantity.

L-ASCORBIC ACID 500GM Catalog #TCA0537-500G

Product TCA0537-500G is a replacement for AAAA15613-18 and has the same packaging and specifications, but may be a different quantity.

L-(+)-ASCORBIC ACID 99% 100G Catalog #BT218855-100G

Product BT218855-100G is a replacement for AAAA15613-18 and has the same packaging and specifications, but may be a different quantity.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99+% CAS No.: 50-81-7 Molecular Formula: C6H8O6 Formula Weight: 176.12 Melting Point: 190-193° dec. Beilstein Reference: 84272 Merck Reference: 14,830 Optical Rotation: +21° (c=10 in water) MDL No.: MFCD00064328

Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:14,830
  • Boiling Pt:553 °C (1013 hPa)
  • Melting Pt:190…193 °C (dec.)
  • Density:1.65
  • Storage Temperature:Ambient
  • EINECS:200-066-2
  • REACH:exempt

Documents

Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    50 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.