Boron nitride, paste
Product Details & Documents
Resistance against molten metals and salts, high thermal conductivity and dielectric strength
Physical Form: Paste Density: 1.40 MDL No.: MFCD00011317
Resistance against molten metals and salts, high thermal conductivity and dielectric strength
- CAS Number:10043-11-5
- Formula:BN
- MDL Number:MFCD00011317
- Molecular Weight:24.82 g/mol
- Merck Index:13,01336
- Melting Pt:2527 °C
- Density:1.40
- Storage Temperature:Ambient
- EINECS:233-136-6
Documents
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



