Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Boron nitride, paste

Boron nitride, paste

bottle, 500 g
Catalog # AAA43773-36
Irritant
Supplier #:  043773-36
Boron nitride, paste
Catalog # AAA43773-36
CAS Number:  10043-11-5
Boron nitride, paste

bottle, 500 g

CAS Number:  10043-11-5
Price
$684.66
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Resistance against molten metals and salts, high thermal conductivity and dielectric strength

Physical Form: Paste Density: 1.40 MDL No.: MFCD00011317

Resistance against molten metals and salts, high thermal conductivity and dielectric strength

  • CAS Number:10043-11-5
  • Formula:BN
  • MDL Number:MFCD00011317
  • Molecular Weight:24.82 g/mol
  • Merck Index:13,01336
  • Melting Pt:2527 °C
  • Density:1.40
  • Storage Temperature:Ambient
  • EINECS:233-136-6
Specifications

Specifications

  • Pack type:
    bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.