Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
D-(+)-Biotin 98+%

D-(+)-Biotin 98+%

Bottle, 1 g
Catalog # AAA14207-03
Supplier #:  A14207-03
D-(+)-Biotin 98+%
Catalog # AAA14207-03
CAS Number:  58-85-5
D-(+)-Biotin 98+%
D-(+)-Biotin 98+%

Bottle, 1 g

CAS Number:  58-85-5
Price
$129.18
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Essential vitamin that is important for amino acid and energy metabolism

Purity: 98+% CAS No.: 58-85-5 Molecular Formula: C10H16N2O3S Formula Weight: 244.32 Storage Temperature: 2°C to 8°C Melting Point: 224-226° Beilstein Reference: 86838 Merck Reference: 14,1231 Optical Rotation: +91° (c=1 in 0.1N NaOH) MDL No.: MFCD00005541

Essential vitamin that is important for amino acid and energy metabolism

Synonyms:  Vitamin H, D-Biotin, Vitamin B8, 5-((3aS,4S,6aR)-2-Oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl)pentanoic acid, Vitamin B7
  • CAS Number:58-85-5
  • Formula:C₁₀H₁₆N₂O₃S
  • MDL Number:MFCD00005541
  • Molecular Weight:244.31 g/mol
  • Merck Index:14,1231
  • Melting Pt:224…226 °C
  • Density:1.41 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-399-3
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.