Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Vanillin 99%

Bottle, 500 g
Catalog # AAA11169-36
Irritant
 :  A11169-36
Vanillin 99%
Catalog # AAA11169-36
CAS Number:  121-33-5
Vanillin 99%
Vanillin 99%

Bottle, 500 g

CAS Number:  121-33-5
Price
$140.41
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00006942 Beilstein Registry No.: 472792

Purity: 99% CAS No.: 121-33-5 Molecular Formula: C8H8O3 Formula Weight: 152.15 Flash Point: 153°(307°F) Physical Form: Crystalline powder Density: 1.06 Sensitivity: Air and Light Sensitive Appearance: White Melting Point: 81-83° Boiling Point: 170°/15mm Beilstein Reference: 472792 Merck Reference: 14,9932 MDL No.: MFCD00006942

Synonyms:  4-Hydroxy-3-methoxybenzaldehyde
  • CAS Number:121-33-5
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00006942
  • Molecular Weight:152.15 g/mol
  • Merck Index:14,9932
  • Boiling Pt:170 °C/15 mm
  • Melting Pt:81…83 °C
  • Density:1.06
  • Flash Pt:153 °C(307 °F)
  • Storage Temperature:Ambient
  • EINECS:204-465-2
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.