Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hydroxylammonium sulphate ≥99%

Hydroxylammonium sulphate ≥99%

Bottle, 2 kg
Catalog # AA88944-A3
Health
Corrosive
Irritant
Environment
Supplier #:  088944-A3
Hydroxylammonium sulphate ≥99%
Catalog # AA88944-A3
CAS Number:  10039-54-0
Hydroxylammonium sulphate ≥99%

Bottle, 2 kg

CAS Number:  10039-54-0
Price
$193.53
Log in for availability
Each (2kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00044869 Soluble in water

Purity: 99% CAS No.: 10039-54-0 Molecular Formula: H3NO.0.5H2SO4 Formula Weight: 164.14 Physical Form: Crystalline Density: 1.88 Appearance: White Odor: Odorless pH: 3.6 Melting Point: ca 170° dec. Solubility: Soluble in water Merck Reference: 14,4828 UN No.: 2865 MDL No.: MFCD00044869

Synonyms:  Bis(hydroxylammonium) sulphate, Hydroxylamine sulphate, Hydroxylamine sulphate (2:1)
  • CAS Number:10039-54-0
  • Formula:H₈N₂O₆S
  • MDL Number:MFCD00044869
  • Molecular Weight:164.14 g/mol
  • Merck Index:14,4828
  • Melting Pt:∼ 170 °C (dec.)
  • Density:1.88
  • Storage Temperature:Ambient
  • EINECS:233-118-8
  • UN:2865
  • ADR:8,III

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    2 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.