Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Lead(II) oxide low silver

Lead(II) oxide low silver

Bottle, 2 kg
Catalog # AA33330-A3
Health
Irritant
Environment
Supplier #:  033330-A3
Lead(II) oxide low silver
Catalog # AA33330-A3
CAS Number:  1317-36-8
Lead(II) oxide low silver

Bottle, 2 kg

CAS Number:  1317-36-8
Price
$90.46
Log in for availability
Each (2kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores

CAS No.: 1317-36-8 Molecular Formula: PbO Formula Weight: 223.19 Physical Form: Powder Density: 9.53 Odor: Odorless Merck Reference: 14,5413 UN No.: 3077 MDL No.: MFCD00011164

As storage batteries, ceramic cements and fluxes, pottery and glazes, in chromium pigments, in oil refining, in paints, and in the assay of precious metal ores

Synonyms:  Lead monoxide
  • CAS Number:1317-36-8
  • Formula:PbO
  • MDL Number:MFCD00011164
  • Molecular Weight:223.2 g/mol
  • Merck Index:14,5413
  • Boiling Pt:1470 °C (1013 hPa)
  • Melting Pt:886 °C
  • Density:9.53
  • Storage Temperature:Ambient
  • EINECS:215-267-0
  • UN:3077
  • ADR:9,III

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    2 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.