Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Silicon dioxide ≥99.5%

Silicon dioxide ≥99.5%

Bottle, 2 kg
  AA13024-A3
 
Supplier #:  013024-A3
Silicon dioxide ≥99.5%
  AA13024-A3
 :  7631-86-9
Silicon dioxide ≥99.5%

Bottle, 2 kg

 :  7631-86-9
 
$77.58
   
Each (2kg)
 
   
 

In the manufacture of glass, ceramics and abrasives, in water filtration, in cosmetics, pharmaceuticals and rubber, as a thermal insulator.

Silica is used in the manufacture of glass for windows, drinking glasses, beverage bottles, and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware, and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

98.5-99% microcrystalline silica in the form of quartzIt is a very stable compound of silicon.

 :  Silicon(IV) oxide, Silicium dioxide, Silicium (IV) oxide
  •  :7631-86-9
  •  :SiO₂
  •  :MFCD00011232
  •  :60.08 g/mol
  •  :14,8493
  •  :2230 °C
  •  :1710 °C
  •  :2.2 g/cm³ (20 °C)
  •  :Ambient
  •  :238-878-4
 

 

  • Pack type:
    Bottle
  • Size:
    2 kg

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.