Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Vanadium(III) oxide ≥95%

Bottle, 250 g
Catalog # AA12301-30
Toxic
Irritant
 :  012301-30
Vanadium(III) oxide ≥95%
Catalog # AA12301-30
CAS Number:  1314-34-7
Vanadium(III) oxide ≥95%

Bottle, 250 g

CAS Number:  1314-34-7
Price
$102.42
Log in for availability
Each (250g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

MDL: MFCD00011455 Insoluble in water

Purity: 95% CAS No.: 1314-34-7 Molecular Formula: O3V2 Formula Weight: 149.88 Physical Form: Powder Density: 4.87 Appearance: Black Odor: Odorless Melting Point: 1970° Solubility: Insoluble in water Merck Reference: 14,9923 UN No.: 3285 MDL No.: MFCD00011455

  • CAS Number:1314-34-7
  • Formula:O₃V₂
  • MDL Number:MFCD00011455
  • Molecular Weight:149.88
  • Merck Index:14,9923
  • Melting Pt:1970 °C
  • Density:4.87
  • Storage Temperature:Ambient
  • EINECS:215-230-9
  • UN:3285
  • ADR:6.1,II
  •  :Density: 4.87 g/cm³ (20 °C)
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.