Orders LinkContactUsLinkComponent
Conductivity Standards, Sachets
Conductivity Standards, Sachets
500 ml, 12.88 mS/cm, Bottles
Catalog # 97007-754
Conductivity Standards, Sachets
Catalog # 97007-754
Supplier:  
CAS Number:  
Conductivity Standards, Sachets
Conductivity Standards, Sachets
500 ml, 12.88 mS/cm, Bottles
CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
STANDARD CONDUCTIVITY 12.88MS 250ML Catalog #76359-840
Product 76359-840 is similar to 97007-754 but manufacturer and specifications may vary.
Product Details & Documents
NIST-traceable standard for conductivity electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Mettler Toledo sachets ensure a fresh portion of standard is used for each calibration, improving your conductivity and TDS data.
Specifications
Specifications
  • Pack type:
    Bottles
  • Size:
    500 ml
  • Conductivity:
    12.88 mS/cm