Orders LinkContactUsLinkComponent
Electrode Fill Solutions
1M KCl, Bottle, 25 ml, —
  97001-716
 :  51343181
Electrode Fill Solutions
  97001-716
 :  
Electrode Fill Solutions
Electrode Fill Solutions
1M KCl, Bottle, 25 ml, —
 :  
$26.80
Each (25ml)
FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.
 : Supplied in 25 ml (0.9 oz.) bottles.
  • Pack type:
    Bottle
  • Description:
    1M KCl
  • Ion:
  • Volume:
    25 ml