Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

D-(-)-Mannitol

Glass bottle, 500 g
Catalog # 80108-992
Supplier:   MilliporeSigma
 :  443907-500GM
D-(-)-Mannitol
Catalog # 80108-992
Supplier:  MilliporeSigma
CAS Number:  69-65-8
D-(-)-Mannitol

Glass bottle, 500 g

CAS Number:  69-65-8
Price
$267.29
Log in for availability
Each (500g)
 
Product Details & Documents
Synonyms:  Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol
  • CAS Number:69-65-8
  • Formula:HOH₂C(CH(OH))₄CH₂OH
  • MDL Number:MFCD00064287
  • Molecular Weight:182.17 g/mol

 

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Size:
    500 g

Specifications