Inoculation Loop, Globe Scientific
Inoculation Loop, 1 uL and 10 uL, White, Sterile, 10/Peel Pack, 50 Packs/Case
Catalog # 60820-132
Supplier: Globe Scientific
Supplier #: 2860
Jump to:
Product Details & Documents
Loops offer the most convenient and safest way of streaking culture media
- Polystyrene (PS)
- Certificate of Calibration comes with each case of product
- Gamma sterilized
- Rigid feel for comfortable, stable, uniform streaking
- One side is a 10 µl loop and the reverse side is a 1 µl loop
Loops are sterilized by gamma radiation.
Molded from a special medical grade polystyrene, they offer a strong, rigid feel to provide comfortable, stable, and uniform streaking.
Delivery information: Supplied with calibration certificate.
Documents
Certificates
Search by Lot NumberSpecifications
- Sterility:Sterile
- Color:White
- Description:Inoculation Loop, 1 uL and 10 uL
- Packaging:10/Peel Pack, 50 Packs/Case