Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GLPC ACID CLEAN COMPOUND 15GL
Catalog # 21840-134
Supplier:
Quip Laboratories
SDS
Expand image
GLPC ACID CLEAN COMPOUND 15GL
Catalog # 21840-134
Supplier:
Quip Laboratories
Supplier Number:
GLPAC-15GAL
SDS
Related Information
Documentation
SDS
ghs_GLPAC.pdf
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation