Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLVENT SWITCH FS-21 240V
Catalog # 21506-127
Supplier:
Metrohm USA
Expand image
SOLVENT SWITCH FS-21 240V
Catalog # 21506-127
Supplier:
Metrohm USA
Supplier Number:
022853139