$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
DECONSPORE .06/16Z N/S
14003-720
:
Veltek Associates
DECONSPORE .06/16Z N/S
14003-720
:
Veltek Associates
:
DS200-0716Z2
ghs_DS200-01A
VWR GHS Compliance Letter (US)