$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CAL GAS SO2 10PPM 58L IN N2
13453-510
:
Draeger
CAL GAS SO2 10PPM 58L IN N2
13453-510
:
Draeger
:
4597050
ghs_4597050.pdf
VWR GHS Compliance Letter (US)