CTAB Extraction Solution
CTAB extraction solution, sterile, 250 ml
Jump to:
Discontinued
Product Details & Documents
CTAB is a cationic detergent used in the extraction of genomic DNA from bacteria and for the isolation of high molecular weight DNA from plants for plamid minipreps and for PCR.
The Solution contains 1% CTAB, 100 mM Tris-HCl (pH 8,0), 20 mM EDTA (pH 8,0) and 1.4 M Sodium Chloride.
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Description:CTAB extraction solution, sterile
- Size:250 ml



