Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Galactose Solution, Teknova

Galactose Solution, Teknova

Galactose Solution, 20%, 1000 mL
Catalog # 100217-128
Galactose Solution, Teknova
Catalog # 100217-128
Supplier:  
CAS Number:  
Galactose Solution, Teknova

Galactose Solution, 20%, 1000 mL

CAS Number:  
Discontinued
Product Details & Documents

For used in growth media and stored at room temperature.

Specifications

Specifications

  • Description:
    Galactose Solution, 20%
  • Size:
    1000 mL
  • Cat. No.:
    100217-128
  • Sterility:
    Sterile

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.