Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Pierce™ Wash Buffers, Thermo Scientific

Pierce™ Wash Buffers, Thermo Scientific

Pierce™ TBS Tween™ 20 Buffer, 20X, 500 mL
Catalog # PI28360
Marketsource
Supplier:   Invitrogen
Supplier #:  28360
Pierce™ Wash Buffers, Thermo Scientific
Catalog # PI28360
Supplier:  Invitrogen
Marketsource
CAS Number:  
Pierce™ Wash Buffers, Thermo Scientific
Pierce™ Wash Buffers, Thermo Scientific

Pierce™ TBS Tween™ 20 Buffer, 20X, 500 mL

CAS Number:  
Price
$158.77
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

These wash buffers are designed for use in dialysis, cross-linking, enzyme assays, ELISAs, immunohistochemistry, protein plate-coating, biotinylation applications.

  • Ready to use without having to reconstitute with ultrapure water
  • Eliminates the possibility of powder remaining in a packet
  • 10X or 20X concentration eliminates time spent waiting for powder to dissolve
  • Storage as concentrated stock minimizes bench space needed for large volume solutions

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 20 to 25˚C.

Caution: For Research Use Only. Not for use in diagnostic procedures.

Specifications

Specifications

  • Description:
    Pierce™ TBS Tween™ 20 Buffer, 20X
  • Size:
    500 mL
  • Cat. No.:
    PI28360

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.