Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB), powder

3,3',5,5'-Tetramethylbenzidine (TMB), powder

5 g
Catalog # IC980501
Supplier:   MP Biomedicals
Supplier #:  08980501
3,3',5,5'-Tetramethylbenzidine (TMB), powder
Catalog # IC980501
Supplier:  MP Biomedicals
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB), powder
3,3',5,5'-Tetramethylbenzidine (TMB), powder

5 g

CAS Number:  54827-17-7
Price
$297.00
Log in for availability
Each (5g)
Minimum order quantity is 1
Product Details & Documents

(TMB Powder) This product is excellent for ELISA procedures.

Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
Specifications

Specifications

  • Size:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.