Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ethidium bromide, powder

Ethidium bromide, powder

1 g
Catalog # IC19020201
Toxic
Health
Supplier:   MP Biomedicals
Supplier #:  0219020201
Ethidium bromide, powder
Catalog # IC19020201
Supplier:  MP Biomedicals
CAS Number:  1239-45-8
Ethidium bromide, powder

1 g

CAS Number:  1239-45-8
Price
$39.50
Log in for availability
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ethidium bromide (EtBr) is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis.

The fluorescence of EtBr increases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the background is not necessary with a low stain concentration (10 µg/ml). Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids. It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA. Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase. Also acts as an RNA polymerase inhibitor, and in separation of high molecular weight DNA's. Confers a deep red stain to DNA by acting as an intercalating agent between the starch bases. Subsequent fluorescence techniques enable the extraction and characterisation of DNA bands.

Ethidium bromide intercalates double-stranded DNA and RNA and acts as a frameshift mutagen. It can also be used in conjunction with acridine orange to differentiate between viable, apoptotic and necrotic cells.

Soluble in water (10mg/mL), Slightly soluble in ethanol; partially soluble in chloroform. Ethidium bromide can also be incorporated into the gel and running buffer at 0.5 µg/mL and visualized immediately after electrophoresis.

Fluorescence: λex 480 nm; λem 620 nm in H2O; λex 530 nm; λem 600 nm in 50 mM phosphate buffer pH 7.0 (upon binding to DNA).

Ethidium bromide (EtBr) is the most commonly used nucleic acid stain for PAGE or agarose gel electrophoresis. The fluorescence of EtBr increases 21-fold upon binding to double-stranded RNA and 25-fold on binding double-stranded DNA so that destaining the background is not necessary with a low stain concentration (10 µg/mL). Ethidium bromide has been used in a number of fluorimetric assays for nucleic acids. It has been shown to bind to single-stranded DNA (although not as strongly) and triple-stranded DNA. Because of its ability to bind to DNA, EtBr is an inhibitor of DNA polymerase. Also acts as an RNA polymerase inhibitor, and in separation of high molecular weight DNA's. Confers a deep red stain to DNA by acting as an intercalating agent between the starch bases. Subsequent fluorescence techniques enable the extraction and characterization of DNA bands.

Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Melting Pt:260…262 °C (lit.)
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,I
Specifications

Specifications

  • Size:
    1 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.