Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Rapamycin ≥98%

Rapamycin ≥98%

100 µg
Catalog # IC15934680
Supplier:   MP Biomedicals
Supplier #:  0215934680
Rapamycin ≥98%
Catalog # IC15934680
Supplier:  MP Biomedicals
CAS Number:  53123-88-9
Rapamycin ≥98%

100 µg

CAS Number:  53123-88-9
Price
$142.00
Log in for availability
Each (100µG)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Rapamycin is a member of the macrolide immunosuppressant family and a FRAP inhibitor and inhibits lymphokine induced cell proliferation at the G1 and S phase as well as an irreversible cellular arrest at the G1 phase in S. cerevisiae cells.

  • White powder

This antibiotic also exhibits selective signal blocking leading to the activation of p70/85 S6 kinase, which is potentially due to the inhibition of FRAP autophosphorylation or protein kinase activity

This rapamycin binds to and inhibits the molecular target of rapamycin (mTOR). It forms a complex with FKBP12 that binds to and inhibits the molecular target of rapamycin (mTOR). Rapamycin is a potent immunosuppressant and has anticancer activity.

Angiogenesis inhibition is also exhibited, possibly through the inhibition of the Akt pathway. It also induces autophagy in yeast and mammalian cell lines.

Extinction coefficients: E1 %, 1 cm (maximum wavelength, 95% ethanol): 417 (267 nm); 541 (277 nm); 416 (288 nm)

Soluble in ether, chloroform, acetone,DMF, methanol (25 mg/mL - clear, colorless solution) or DMSO, very sparingly soluble in hexane and petr ether and insoluble in water

Synonyms:  (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
  • CAS Number:53123-88-9
  • Formula:C₅₁H₇₉NO₁₃
  • MDL Number:MFCD00867594
  • Molecular Weight:914.19 g/mol
  • Storage Temperature:Freezer

Documents

Specifications

Specifications

  • Size:
    100 µg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.