L(+)-Ascorbic acid ≥98%
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
L-Ascorbic Acid is used as an Antimicrobial and Antioxidant in foodstuffs. Reductant for the isolation of chloroplasts. It has an effect on the nonspecific immune system. Preserves nitric oxide (NO) which may help prevent endothelial dysfunction. It also helps prevent and reverse the oxidative damage done to proteins and lipids as a result of cigarette smoke.
L-Ascorbic Acid, free Acid protects cells against the damaging effects of radiation and oxygen radicals. L-Ascorbic Acid increases the rate of mineralization in osteoblasts. L-Ascorbic Acid is involved in hydroxylation of proline and lysine. L-Ascorbic Acid is a calcium channel protein inhibitor and reduces Na+/Ca2+ exchange in cultured astrocytes. L-Ascorbic Acid modulates cyclic nucleotide levels in B and T cells inhibits apoptosis in cultured rat ovarian follicles.
Soluble in alcohol, water (1 g/3 mL; 80% w/v at 100 °C; 40% w/v at 45 °C), absolute ethanol (1 g/50 mL), glycerol (1 g/100 mL), or propylene glycol (1 g/20 mL); insoluble in ether, chloroform, benzene, petroleum ether, oils, fats, fat solvents.
Aqueous solutions are rapidly oxidized by air; the reaction is accelerated by alkalies, iron, and copper.
- CAS Number:50-81-7
- Formula:C₆H₈O₆
- MDL Number:MFCD00064328
- Molecular Weight:176.13 g/mol
Documents
VWR GHS Compliance Letter (US)
Additional Product Details
Specifications
Specification Test Results
- Synonyms:Vitamin C; Ascorbate; Sodium ascorbate; L-Xyloascorbic acid; 3-Oxo-L- gulofuranolactone (enol form); L-3-Ketothreohexuronic acid lactone; Antiscorbutic vitamin; Cevitamic acid; Sodascorbate
- Form:White to slightly yellow crystalline powder
- Storage:Store at Room Temperature .Protect from light
- Optical Rotation:+18° to +32° (c=1, water)
- pH:1 - 3 (10% aq solution)
- pka:pK1: 4.17; pK2: 11.57
Specifications
- Size:1 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



