Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
PBS Buffer, Cytiva

PBS Buffer, Cytiva

PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl, 1000 ml
Catalog # 101724-758
Marketsource
Supplier:   Cytiva
Supplier #:  BR100672
PBS Buffer, Cytiva
Catalog # 101724-758
Supplier:  Cytiva
Marketsource
CAS Number:  
PBS Buffer, Cytiva
PBS Buffer, Cytiva

PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl, 1000 ml

CAS Number:  
Price
$337.68
Log in for availability
Each
This product has been blocked by your organization. Please contact your purchasing department for more information
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Phosphate buffered saline solution containing detergent, which is recommended as running buffer for small molecule analysis in Biacore systems. Concentrated stock solution, yielding pH 7,4 when diluted 10X.

  • Can be stored at room temperature, precipitation can occur at low temperature
  • Supplemented with DMSO
Specifications

Specifications

  • Description:
    PBS buffer 10X, includes 0,1 M phosphate buffer with 27 mM KCl and 1,37 M NaCl
  • Size:
    1000 ml
  • Cat. No.:
    101724-758
  • For Use With:
    Interaction analyses in Biacore systems, most commonly small molecule assays

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.