Orders LinkContactUsLinkComponent
Modular Blocks, Electron Microscopy Sciences
Modular Blocks, Electron Microscopy Sciences
Modular block, 2065Q, Titer Plate, size of two blocks, 1
Catalog # 101412-766
Supplier #:  63133-02
Modular Blocks, Electron Microscopy Sciences
Catalog # 101412-766
Modular Blocks, Electron Microscopy Sciences
Modular Blocks, Electron Microscopy Sciences
Modular block, 2065Q, Titer Plate, size of two blocks, 1
Price
$181.40
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Solid black anodized aluminum blocks for better heat retention.
Block includes thermometer well for monitoring block temperatures. Close contact of tubes to block walls, provides excellent heat transfer. Block puller included with all models. For use with all our brand modular dri-baths. Single block size: 3.75W×2.9D×2" H (9.5×7.4×5.1 cm).
Specifications
  • Model:
    2065Q
  • Description:
    Modular block
  • No. of wells:
    1
  • Holds:
    Titer Plate, size of two blocks
Accessories
Product Information
  • Catalog #
    Model
    Holds
    Availability
    Price
  • Stainless steel cover for models 2064Q and 2065Q
    Stainless steel cover for models 2064Q and 2065Q
    76421-774
    Log in for availability
    Each - $135.97