Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
DOT Labels 'Explosive Class 1.4 C', Labelmaster

DOT Labels 'Explosive Class 1.4 C', Labelmaster

Preprinted dot labels, Permanent vinyl, Explosive 1.4 C, 100×100 mm (3.94×3.94"), 11.2 mil., On roll
Catalog # 101378-008
Marketsource
Supplier:   Label Master
Supplier #:  SLEXP14C
DOT Labels 'Explosive Class 1.4 C', Labelmaster
Catalog # 101378-008
Supplier:  Label Master
Marketsource
DOT Labels 'Explosive Class 1.4 C', Labelmaster

Preprinted dot labels, Permanent vinyl, Explosive 1.4 C, 100×100 mm (3.94×3.94"), 11.2 mil., On roll

Price
$130.25
Log in for availability
Pack of 500
Minimum order quantity is 1
Product Details & Documents

Help stay in compliance with standards set by the DOT with Explosive Class 1.4 C labels, revised to meet the design requirements of 49 CFR 172.407.

  • Printed with light-fast inks for fade resistance
  • Adhesives formulated for use in various climates and environments
  • Strong holds on corrugated boxes
  • Once affixed, labels won't fade or fall off
  • Outstanding chemcial and abrasion resistance

These labels also meet the design and durability standards of 49 CFR, ICAO, and other international air regulations.

Specifications
  • Description:
    Preprinted dot labels
  • Dimensions:
    100×100 mm (3.94×3.94")
  • Material:
    Permanent vinyl
  • Thickness:
    11.2 mil.
  • Packaging:
    On roll
  • Legend:
    Explosive 1.4 C