Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Phosphate buffers for microscopy staining, Electron Microscopy Sciences

Phosphate buffers for microscopy staining, Electron Microscopy Sciences

Phosphate Buffer, 7.0, 500 ml
Catalog # 100504-554
Supplier #:  26156-02
Phosphate buffers for microscopy staining, Electron Microscopy Sciences
Catalog # 100504-554
CAS Number:  
Phosphate buffers for microscopy staining, Electron Microscopy Sciences

Phosphate Buffer, 7.0, 500 ml

CAS Number:  
Price
$47.04
Log in for availability
Each (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

pH 6.5 is typically used with Wrights stain and potassium ferricyanide solution for Prussian Blue staining of non hemoglobin iron by method of Sundberg and Broman (1955). pH 7.0 buffer is used with Giemsa stain.

Specifications

Specifications

  • Description:
    Phosphate Buffer
  • Size:
    500 ml
  • pH:
    7.0

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.