Orders LinkContactUsLinkComponent
Film Marker, Electron Microscopy Sciences
Film Marker, Electron Microscopy Sciences
Film Marker with 8x Magnifier
Catalog # 100502-794
Supplier #:  68127
Film Marker, Electron Microscopy Sciences
Catalog # 100502-794
Film Marker, Electron Microscopy Sciences
Film Marker, Electron Microscopy Sciences
Film Marker with 8x Magnifier
Price
$92.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This device when fitted to the 8X magnifier (68125) enables you to view and check the 35mm roll film while marking the desired frame by depressing the puncher (located on the side of the device). The puncher punches out a V-shaped piece of film on the perforation to mark the frame. When fitted in the reverse position to the 8X magnifier, you are able to view and check the slides.
Specifications
  • Description:
    Film Marker with 8x Magnifier