Orders LinkContactUsLinkComponent
All Plastic Syringes, Electron Microscopy Sciences
All Plastic Syringes, Electron Microscopy Sciences
Syringe 10 cc, Clear, Eccentric
Catalog # 100500-892
Supplier #:  72524
All Plastic Syringes, Electron Microscopy Sciences
Catalog # 100500-892
All Plastic Syringes, Electron Microscopy Sciences
All Plastic Syringes, Electron Microscopy Sciences
Syringe 10 cc, Clear, Eccentric
Price
$75.21
Log in for availability
Box of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All plastic syringes are the choice for any situation needing an inert, hypoallergenic syringe. Because of their purity, they are indicated for IVF, embryo transfer, chromatography and many laboratory procedures.
  • Latex free and contain no rubber, no silicone oil or styrene
  • Manufactured only from laboratory grade polypropylene and polyethylene
  • Unique plastic syringes have a positive safety stop to prevent accidental spills
 : These syringes are individually sterile strip packed.
Specifications
  • Type:
    Eccentric
  • Color:
    Clear
  • Description:
    Syringe 10 cc