Orders LinkContactUsLinkComponent
Square and Grid Scales, Electron Microscopy Sciences
Square and Grid Scales, Electron Microscopy Sciences
NE10 Graticule, 21 mm, Grid (net) 0.5 mm pitch
Catalog # 100499-150
Supplier #:  68011-21
Square and Grid Scales, Electron Microscopy Sciences
Catalog # 100499-150
Square and Grid Scales, Electron Microscopy Sciences
Square and Grid Scales, Electron Microscopy Sciences
NE10 Graticule, 21 mm, Grid (net) 0.5 mm pitch
Price
$227.91
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Simple grids are convenient for making sketches of the obseved speimen on graph paper.
They are also useful for particle counting.
Specifications
  • Description:
    NE10 Graticule
  • Size:
    21 mm
  • Divisions:
    Grid (net) 0.5 mm pitch