Glennel®, Diamond Suspension, Electron Microscopy Sciences
Product Details & Documents
Glennel® Diamond Suspension assures a consistent, uniform viscosity and dispersion of diamond without any shaking or stirring.
- A versatile, water based suspension designed for most production applications
- Excellent lubrication
- Uniform fast lapping action
- Maximum cutting action
- Compatible with all common extenders
The proprietary formulation guarantees that the particles will stay suspended indefinitely. It is ideal for automatic polishers and applications that require changing operators.
: All Diamond Suspensions are packed in 400 g bottles and have Medium Concentration prepared with Premium Polycrystalline Diamond in either Propylene Glycol (Oil based) or Water based formulas.
Specifications
- Size:9.0 µm