Orders LinkContactUsLinkComponent
Negative Carriers, Electron Microscopy Sciences
Mounted 35 mm Slide
  100494-084
Negative Carriers, Electron Microscopy Sciences
  100494-084
 :  
Negative Carriers, Electron Microscopy Sciences
Negative Carriers, Electron Microscopy Sciences
Mounted 35 mm Slide
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts
Glassless, sandwich type negative carriers are rofessional quality, with black, anodized aluminum construction. Hinged for quick handling.
Universal glass-type masking negative carriers features four individually adjustable masking blades and removable glass inserts. Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
Accepts accessory anti-Newton glass. Top and bottom glass can be removed to convert it to a single-size glassless carrier by means of lock-in inserts.
  • Description:
    Mounted 35 mm Slide