Orders LinkContactUsLinkComponent
Camera Length, Electron Microscopy Sciences
Camera Length, Electron Microscopy Sciences
Evaporated Aluminum Film
Catalog # 100492-366
Supplier #:  80044
Camera Length, Electron Microscopy Sciences
Catalog # 100492-366
Camera Length, Electron Microscopy Sciences
Camera Length, Electron Microscopy Sciences
Evaporated Aluminum Film
Price
$78.10
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The nominal value of the effective camera length of an EM perating in the selected area mode is not sufficiently accurate for any calculations of lattice spacing.
The actual value of camera length must be calibrated as the same accelerating voltage and objective lens setting by reference to a known substance with well defined diffraction spacings. The normal specimens are evaporated films on a 3.05 mm grid of aluminum or thallous chloride. The very small crystalline size yields ring patterns suitable for calibration purposes.
Specifications
  • Description:
    Evaporated Aluminum Film