Orders LinkContactUsLinkComponent
Perfect Loop, Electron Microscopy Sciences
Perfect Loop Handle Only
  100491-696
Perfect Loop, Electron Microscopy Sciences
  100491-696
Perfect Loop, Electron Microscopy Sciences
Perfect Loop, Electron Microscopy Sciences
Perfect Loop Handle Only
$111.77
Each
The Perfect loop is used to place your thin sections that have been cut on the ultramicrotome, easily on the grid mesh without creases.
The Perfect loop for light microscopy has outside diameter of 7 mm which is suitable for large sections.
  • Description:
    Perfect Loop Handle Only