Orders LinkContactUsLinkComponent
Tissue Mounting Stubs, Electron Microscopy Sciences
Aluminum Tissue Mounting Stub
  100490-730
 :  70145-05
Tissue Mounting Stubs, Electron Microscopy Sciences
  100490-730
Tissue Mounting Stubs, Electron Microscopy Sciences
Tissue Mounting Stubs, Electron Microscopy Sciences
Aluminum Tissue Mounting Stub
$164.41
Pack of 50
Machined from aluminum rods with one end tapered to 4.8 mm in diameter. The specimen can be oriented and mounted to the tip with glue and it is ready for sectioning. The body size is "00" and it measures 11 mm long.
  • Description:
    Aluminum Tissue Mounting Stub