Dropping Bottles, Glass, Electron Microscopy Sciences
Product Details & Documents
These bottles deliver stains and indicator solutions drop by drop from a projecting drip tip on the side of the stopper.
A partial turn of the ground glass stopper will seal the bottle.
Specifications
- Description:Amber Dropping Bottle, 100 ml