Orders LinkContactUsLinkComponent
Anionic Gold Tracer, Electron Microscopy Sciences
EM Anionic Gold Tracer, 5 ml, UltraSmall
Catalog # 100487-884
 :  25461
Anionic Gold Tracer, Electron Microscopy Sciences
Catalog # 100487-884
CAS Number:  
Anionic Gold Tracer, Electron Microscopy Sciences
Anionic Gold Tracer, Electron Microscopy Sciences
EM Anionic Gold Tracer, 5 ml, UltraSmall
CAS Number:  
Price
$289.99
Log in for availability
Each (5ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Anionic tracers are used to visualize positive charged moieties in specimens
  • Particle sizes of 0 8, 6, 10, 15 and 25 nm
  • Guaranteed shelf life of 18 months
  • Supplied in PBS with 15 mM NaN3
Aurion Gold Tracers are used to visualize charged moieties in specimens: anionic tracers bind to polycationic moieties (basic proteins e.g. histones), cationic tracers bind to polyanionic moieties (membranes, acidic proteins).
Specifications
Specifications
  • Description:
    EM Anionic Gold Tracer
  • Size:
    UltraSmall
  • Volume:
    5 ml